To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Serum Response Factor
- Clonality:Polyclonal
- Gene ID:6722
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Bovine,Dog,Mouse
- Antigen symbol:SRF
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Serum response Factor (c-fos Serum Response Element-binding transcription Factor|MCM1
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5mg/ml
- Immunogen:C-QASPSRDSSTDLTQ
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89296-308
- Supplier No.:GTX88384
Specifications
About this item
Goat polyclonal antibody to SRF
Approx 67kDa band observed in primary cultured Human Lung fibroblasts lysates after 24 h treatment with 1ng/ml TGF-bl (calculated MW of 51.6kDa according to NP_003122.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Davis FJ et al, Am J Physiol Heart Circ Physiol. 2002 Apr;282(4):H1521-33.; PMID: 11893590). Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: SRF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog