To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cadherin 13
- Clonality:Polyclonal
- Gene ID:1012
- Host:Rabbit
- Reactivity:Human,Mouse
- Antigen symbol:CDH13
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:CDH13|T-Cadherin|Heart Cadherin|CDHH|P105|H-Cadherin|T-cad|Cadherin-13|Truncated Cadherin
- Storage buffer:PBS, 0.02% sodium azide.
- Storage temperature:Store at 4°C, stable for one year.
- Concentration:1mg/ml
- Immunogen:Synthetic peptide - KLH conjugated
- Purification:This antibody was purified using Immunoaffinity Chromatography
- Cat. no.:89271-490
- Supplier No.:GTX85891
Specifications
About this item
Rabbit polyclonal antibody to CDH13
WB (0.5 - 1 µg/ml), IF, IHC-P (5 µg/ml)
Type: Primary
Antigen: CDH13 (Cadherin 13 H-Cadherin (heart))
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: