To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Synaptotagmin 1
- Clonality:Polyclonal
- Gene ID:6857
- Host:Chicken
- Isotype:IgY
- Reactivity:Human,Mouse
- Antigen symbol:SYT1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:Syt1|Synaptotagmin-2|Synaptotagmin II|p65|SYT2|FLJ42519|SytI|Synaptotagmin-1|synaptotagmin 2|Synaptotagmin I|SytII
- Storage buffer:PBS + sodium azide (0.02%, w/v)
- Storage temperature:4°C in the dark
- Concentration:200µg/ml
- Immunogen:Peptides
- Purification:Antigen affinity purification
- Cat. no.:89337-116
- Supplier No.:GTX85467
Specifications
About this item
Chicken polyclonal antibody to Synaptotagmin (Neuron cell marker)
Recommended starting dilutions: For IHC/IF: Use at 1:500 - 1:1000, for WB: Use at 1:1000 - 1:2500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: SYT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse