Anti-PLXDC2 Rabbit Polyclonal Antibody
Catalog #: 89329-490
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Plexin Domain-Containing Protein 2
- Clonality:Polyclonal
- Gene ID:84898
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:PLXDC2
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Plexin domain-containing protein 2|TEM7R|Tumor endothelial marker 7-related protein|UNQ2514/PRO6003
- Storage buffer:PBS containing 0.02% sodium azide
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Purification:Affinity chromatography purified via peptide column
- Cat. no.:89329-490
- Supplier No.:GTX85420
Specifications
About this item
Rabbit polyclonal antibody to Plxdc2
Type: Primary
Antigen: PLXDC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Customers Who Bought This Also Bought