To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B
- Clonality:Monoclonal
- Clone:2C5
- Gene ID:7919
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:BAT1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:D6S81E|BAT1|UAP56
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0.02% sodium azide.
- Storage temperature:For short-term storage, store at 4 °C or store at –20 °C. For long-term storage, store at -70°C (aliquotted). Avoid repeated freeze-thaw cycles.
- Immunogen:Full length human recombinant protein of human BAT1(NP_542165) produced in HEK293T cell.
- Purification:Purified from mouse ascites fluids by Protein A/G affinity chromatography
- Cat. no.:89274-452
- Supplier No.:GTX84837
Specifications
About this item
Mouse monoclonal antibody [2C5] to BAT1
Recommended Starting Dilutions: WB 1:500 IHC 1:50,Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: BAT1
Clonality: Monoclonal
Clone: 2C5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human