To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Beta
- Clonality:Monoclonal
- Clone:BA3R
- Gene ID:60
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Rabbit,Chicken,Human,Rat,Mouse
- Antigen symbol:ACTB
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- Storage buffer:10mM PBS, pH 7.2
- Storage temperature:Store at 4 °C (add 0.1% NaN3) for several days to weeks, and at –20 °C for longer periods. Avoid repeated freeze-thaw cycles
- Concentration:1mg/ml
- Immunogen:β-Actin N-terminal peptide-KLH conjugates
- Purification:Protein A affinity chromatography from mouse ascites fluid
- Cat. no.:89334-110
- Supplier No.:GTX82559
Specifications
About this item
Mouse monoclonal antibody [BA3R] to Actin
Recommended Starting Dilutions:WB (with ECL): Use at a dilution of 1:1000-10000 Immunostaining: Use at a dilution of 1:500 - 1:2000For best results with other assays (e.g.: Dot, ELISA, IP, etc), please determine optimal working dilution by titration test
Type: Primary
Antigen: ACTB
Clonality: Monoclonal
Clone: BA3R
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Chicken, Rabbit, Rat