To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:General Transcription Factor 2H Subunit 4
- Clonality:Polyclonal
- Gene ID:2968
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:GTF2H4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:400 μl
- Western blot:Yes
- Antigen synonyms:TFB2|TFIIH|P52
- Storage buffer:PBS with 0.09% (W/V) sodium azide
- Storage temperature:Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.25mg/ml
- Immunogen:KLH conjugated synthetic peptide between 130~159 amino acids from the Center region of human GTF2H4.
- Purification:Purified through a protein A column, followed by peptide affinity purification.
- Cat. no.:89301-222
- Supplier No.:GTX81063
Specifications
About this item
Rabbit polyclonal to GTF2H4 (Internal)
Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: GTF2H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human