Order Entry
Canada
ContactUsLinkComponent
 
Anti-CASD1 (CAS1 Domain containing 1) Rabbit Polyclonal Antibody
Catalog # 89301-062
Supplier:  Genetex
Anti-CASD1 (CAS1 Domain containing 1) Rabbit Polyclonal Antibody
Catalog # 89301-062
Supplier:  Genetex
Supplier Number:  GTX80980
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CAS1 Domain Containing 1
  • Antigen symbol:
    CASD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Gel shift:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Size:
    400 μl
  • Environmentally Preferable:
  • Gene ID:
    64921
  • Antigen synonyms:
    NBLA04196|C7orf12
  • Storage buffer:
    PBS with 0.09% (W/V) sodium azide
  • Storage temperature:
    Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
  • Concentration:
    0.25mg/ml
  • Immunogen:
    KLH conjugated synthetic peptide selected from the N-terminal region of human CASD1.
  • Purification:
    Purified through a protein A column, followed by peptide affinity purification.
  • Cat. no.:
    89301-062
  • Supplier No.:
    GTX80980

Specifications

About this item

Rabbit polyclonal to CASD1 (N-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CASD1 (CAS1 Domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse