To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:UDP-Gal:betaGlcNAc beta 1 4- galactosyltransferase Polypeptide 1
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:2683
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:B4GALT1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:400 μl
- Western blot:Yes
- Antigen synonyms:beta4Gal-T1|GT1|B4GAL-T1|GGTB2|GTB|CDG2D
- Storage buffer:PBS with 0.09% (W/V) sodium azide
- Storage temperature:Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
- Concentration:0.25mg/ml
- Immunogen:KLH conjugated synthetic peptide between 336~366 amino acids from the C-terminal region of human B4GalT1.
- Purification:Purified through a protein A column, followed by peptide affinity purification.
- Cat. no.:89301-020
- Supplier No.:GTX80958
Specifications
About this item
Rabbit polyclonal to B4GalT1 (C-term)
Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: B4GALT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human