To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of Differentiation 44 antigen
- Clonality:Monoclonal
- Clone:IM7
- Gene ID:960
- Host:Rat
- Isotype:IgG2b
- Reactivity:Horse,Human,Dog,Mouse,Cat
- Antigen symbol:CD44
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:MDU2|HUTCH-I|LHR|MC56|HCAM|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
- Storage buffer:Phosphate buffered saline (PBS) with 15 mM sodium azide, approx. pH 7.4
- Storage temperature:Store in the dark at 2-8°C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:Dexamethasone-induced cells of the SJL mouse spontaneous myeloid leukemia M1
- Cat. no.:89316-938
- Supplier No.:GTX80087
Specifications
About this item
FITC-conjugated Rat Monoclonal antibody [IM7] to CD44
Recommended Starting Dilutions:FACS: Use at 1 µg/mL. Positive Control:mouse peripheral bloodIHC-P: Positive tissue:mouse spleenOptimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: