To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD14 Molecule
- Clonality:Monoclonal
- Clone:MEM-18
- Gel shift:Yes
- Gene ID:929
- Host:Mouse
- Isotype:IgG1
- Reactivity:Primates,Human
- Antigen symbol:CD14
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Size:100 test
- Western blot:Yes
- Antigen synonyms:monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
- Storage buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage temperature:Store in the dark at 2-8°C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:A crude mixture of human urinary proteins precipitated by ammonium sulphate from the urine of a patient suffering from proteinuria.
- Cat. no.:89298-846
- Supplier No.:GTX79990
Specifications
About this item
APC-conjugated Mouse Monoclonal antibody [MEM-18] to CD14
Recommended Starting Dilutions:WB: Use under non-reducing conditionsELISA: The antibody MEM-18 has been tested as the detection antibody in a sandwich ELISA for analysis of human CD14 in combination with antibody B-A8 (cat. no. 11-304-C100)Functional: The antibody MEM-18 completely blocks binding of fluorescein (FITC) –labeled bacterial LPS to the monocyte surface and it also blocks the binding of CD14 to the extracellular TLR2 domainOptimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MEM-18
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Primate