To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Endoglin
- Clonality:Monoclonal
- Clone:MEM-226
- Gel shift:Yes
- Gene ID:2022
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:ENG
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Size:100 test
- Western blot:Yes
- Antigen synonyms:Eng|AI662476|AI528660|CD105|endoglin|END|Endo|ORW1|HHT1|S-endoglin
- Storage buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage temperature:Store in the dark at 2-8°C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:Recombinant <i>Vaccinia</i> virus containing the human CD105 cDNA.
- Cat. no.:89298-832
- Supplier No.:GTX79979
Specifications
About this item
PE-conjugated Mouse Monoclonal antibody [MEM-226] to CD105
Recommended Starting Dilutions:WB: Use under non-reducing conditionsOptimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: MEM-226
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human