To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of Differentiation 44 antigen
- Clonality:Monoclonal
- Clone:MEM-85
- Gel shift:Yes
- Gene ID:960
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:CD44
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Size:100 test
- Western blot:Yes
- Antigen synonyms:MDU2|HUTCH-I|LHR|MC56|HCAM|Pgp1|CDW44|MIC4|ECMR-III|HCELL|MDU3|IN|CSPG8
- Storage buffer:The reagent is provided in phosphate buffered saline (PBS) containing 15 mM sodium azide and 0.2% (w/v) high-grade protease free Bovine Serum Albumin (BSA) as a stabilizing agent.
- Storage temperature:Store in the dark at 2-8°C. Do not freeze. Do not use after expiration date stamped on vial label. Short-term exposure to room temperature should not affect the quality of the reagent.
- Immunogen:Leukocytes of a patient suffering from LGL Type Leukaemia.
- Cat. no.:89298-760
- Supplier No.:GTX79924
Specifications
About this item
PE-conjugated Mouse Monoclonal antibody [MEM-85] to CD44
Recommended Starting Dilutions:WB: Use at 2 µg/mL, 60 min on vertical incubator. Positive Control: Kg-1a human leukemia cell lysate Sample preparation: Resuspend approx. 50 mil. cells in 1 ml cold Lysis buffer (1% laurylmaltoside in 20 mM Tris/Cl, 100 mM NaCl pH 8.2, 50 mM NaF including Protease inhibitor Cocktail). Incubate 60 min on ice. Centrifuge to remove cell debris. Mix lysate with non-reducing SDS-PAGE Sample buffer. Use under non-reducing conditions. SDS-PAGE (6% separating gel)Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: MEM-85
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: