To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 25 (Mitochondrial Carrier; Dicarboxylate Transporter), Member 10
- Clonality:Polyclonal
- Gene ID:27376
- Host:Rabbit
- Reactivity:Mouse
- Antigen symbol:SLC25A10
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:DIC
- Storage buffer:Antisera containing 0.05% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Immunogen:Synthetic peptide corresponding to the C-terminal residues L(276) R K H F G I K V P T T(287) of mDIC.
- Purification:Antiserum
- Cat. no.:89292-332
- Supplier No.:GTX79240
Specifications
About this item
Rabbit Polyclonal antibody to SLC25A10.
Recommended Starting Dilutions: ICC/IF: 1:50 - 1:500 WB: 1:500 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: SLC25A10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: IgG
Host: Rabbit
Isotype:
Reactivity: Mouse