To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD20 molecule
- Clonality:Monoclonal
- Clone:LT20
- Gene ID:931
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:CD20
- Conjugation:Biotin
- Flow cytometry:Yes
- ImmunoFluorescence:Yes
- Size:100 μg
- Antigen synonyms:Membrane-spanning 4-domains subfamily A member 1|CVID5 MS4A2|MS4A1|Bp35|S7|LEU-16|CD20|B1
- Storage buffer:Phosphate buffered saline (PBS) with 15 mM sodium azide, approx. pH 7.4
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Normal human lymphocytes from lymph node.
- Cat. no.:89285-532
- Supplier No.:GTX78378
Specifications
About this item
Monoclonal Antibody to CD20 (Human) Biotin
Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:150. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: LT20
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype:
Reactivity: Human