To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:T-cell Receptor Alpha And Beta
- Clonality:Monoclonal
- Clone:R73
- Gene ID:24820
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Antigen symbol:TCRA +TCRB
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:250 μg
- Antigen synonyms:Tcrb|FLJ22602|TCRA|MGC71411|MGC117436|T-cell receptor|MGC23964|TCRD|TCR beta|MGC22624|alpha polypeptide|TRA|Tcra|T-cell antigen receptor
- Storage buffer:Phosphate buffered saline containing 0.02% sodium azide and BSA
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:Rat T Blasts and Erythrocytes
- Purification:Protein G affinity purified
- Cat. no.:89317-786
- Supplier No.:GTX76146
Specifications
About this item
Mouse monoclonal [R73] to TCR Alpha / Beta
For FACS: Use at an assay dependent dilution. For IHC-Fr: Use at an assay dependent dilution. For IP: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.
Type: Primary
Antigen: TCRA +TCRB
Clonality: Monoclonal
Clone: R73
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat