To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CCT eta
- Clonality:Monoclonal
- Clone:PK/16/8/a
- Gene ID:514355
- Host:Rat
- Isotype:IgG2a
- Reactivity:Rabbit,Human,Mouse
- Antigen symbol:CCT eta
- Conjugation:Unconjugated
- Size:50 μg
- Western blot:Yes
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. no.:89263-508
- Supplier No.:GTX75919
Specifications
About this item
Rat monoclonal antibody [PK/16/8/a] to CCT eta
Recommended Starting Dilutions: Western Blotting: Max Dilution 1/500 Immunohistology - Paraffin: Max Dilution 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CCT eta
Clonality: Monoclonal
Clone: PK/16/81a
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human, Mouse, Rabbit