To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor 5 (gene/pseudogene)
- Clonality:Monoclonal
- Clone:HEK/1/85a
- Gene ID:1234
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:CCR5
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:1 ml
- Antigen synonyms:IDDM22|CKR5|CD195|CMKBR5|CC-CKR-5|CCCKR5|CCR-5|CKR-5
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
- Storage temperature:Store at 4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. no.:89278-754
- Supplier No.:GTX75915
Specifications
About this item
Rat monoclonal antibody [HEK/1/85a] to CCR5
Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: HEK/1/85a
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity: