To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cluster of Differentiation 79a molecule, immunoglobulin-associated beta
- Clonality:Monoclonal
- Clone:HM79-11
- Gene ID:15985
- Host:Hamster
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:CD79B
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Size:500 μg
- Western blot:Yes
- Antigen synonyms:immunoglobulin-associated B29 protein|B29|RP23-418O11.7|Ig-beta|Igbeta|immunoglobulin-associated beta|B-cell-specific glycoprotein B29|B-cell antigen receptor complex-associated protein beta chain|Igb
- Storage buffer:Phosphate buffered saline pH 7.4
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Immunogen:CD79 alpha/CD79 beta heterodimer purified from WEH1231 B cells.
- Purification:Protein G affinity purified
- Cat. no.:89280-806
- Supplier No.:GTX75401
Specifications
About this item
Hamster monoclonal [HM79-11] to Mouse CD79 Beta - Azide Free
For FACS: Use at a dilution of 1:50 - 1:100. For IHC-Fr, IP and WB: Use at an assay dependent dilution. Not tested in other applications. Optimal concentrations/dilutions should be determined by the researcher.
Type: Primary
Antigen: CD79B
Clonality: Monoclonal
Clone: HM79-11
Conjugation:
Epitope:
Host: Hamster
Isotype:
Reactivity: