To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Troponin I Type 3
- Clonality:Monoclonal
- Clone:10F4 (110)
- Gene ID:7137
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Rabbit,Goat,Rat,Cat
- Antigen symbol:TNNI3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:250 μg
- Western blot:Yes
- Antigen synonyms:CMD2A|RCM1|TNNC1|CMD1FF|cTnI|CMH7
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Human Cardiac troponin I
- Purification:Purified IgG prepared by affinity chromatography on Protein A
- Cat. no.:89263-352
- Supplier No.:GTX74745
Specifications
About this item
Mouse monoclonal antibody [10F4 (110)] to Cardiac Troponin I
Type: Primary
Antigen: TNNI3
Clonality: Monoclonal
Clone: 10F4 (110)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Dog, Pig, Rat, Cat, Goat, Rabbit, Bovine