To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) receptor 5 (gene/pseudogene)
- Clonality:Polyclonal
- Gene ID:1234
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CCR5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:IDDM22|CKR5|CD195|CMKBR5|CC-CKR-5|CCCKR5|CCR-5|CKR-5
- Storage buffer:Phosphate buffered saline containing 0.02% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Store undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Peptide corresponding to amino acids 6 to 20 of human CCR5.
- Cat. no.:89278-238
- Supplier No.:GTX74371
Specifications
About this item
Rabbit polyclonal antibody to CCR5
Recommended Starting Dilutions: Western Blotting: Max Dilution 1/1000 Immunohistology - Paraffin: Max Dilution 20 ug/ml Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CCR5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: