To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 16
- Clonality:Monoclonal
- Clone:LL025
- Gene ID:3868
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CK16
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:1 ml
- Antigen synonyms:cytokeratin 16|cytokeratin-16|keratin-16|focal non-epidermolytic palmoplantar keratoderma|CK-16|type I cytoskeletal 16|keratin
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Immunogen:BALB/C mice were injected with a synthetic peptide from the C-terminus of human cytokeratin 16.
- Purification:This antibody is supplied as purified immunoglobulin fraction containing sodium azide as preservative.
- Cat. no.:89277-584
- Supplier No.:GTX73557
Specifications
About this item
Monoclonal Mouse Antibody to Human Cytokeratin 16
This antibody may be diluted to a titer of 1:25-1:75 in an ABC method. However, depending on the staining conditions employed, we suggest that final dilution should be determined by the user. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining.
Type: Primary
Antigen: CK16
Clonality: Monoclonal
Clone: LL025
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: