To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calcitonin-related Polypeptide Alpha
- Clonality:Polyclonal
- Gene ID:796
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:CALCA
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:6 ml
- Antigen synonyms:CT|Calcitonin-related Polypeptide alpha|CGRP|CALC1|KC|CGRP1|CGRP-I|Calcitonin
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C.
- Immunogen:Synthetic human calcitonin 1-32.
- Cat. no.:89315-974
- Supplier No.:GTX73367
Specifications
About this item
Polyclonal Antibody to Human Calcitonin
This antibody has been pretitered and quality controlled to work on formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions should be determined by the user. Suitable for formalin/paraffin tissues.
Type: Primary
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: