To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Prostate Short-chain Dehydrogenase Reductase
- Clonality:Polyclonal
- Gene ID:51109
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:PSDR1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:SDR7C1|RDJCSS|ARSDR1|RALR1|MDT1|HCBP12|PSDR1|CGI82|SCALD
- Storage buffer:PBS containing 0.02% sodium azide.
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Purification:Affinity purified IgG
- Cat. no.:89285-420
- Supplier No.:GTX70423
Specifications
About this item
Rabbit Polyclonal to PSDR1
Western blot - Use at 1:500 to 1:1,000 dilution. However, the investigator should determine the optimal dilution for a specific application such as immunohistochemistry.
Type: Primary
Antigen: RDH11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human