To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Nitric Oxide Synthase 1 (Neuronal)
- Clonality:Polyclonal
- Gene ID:4842
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NNOS
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:bNOS1|Neuronal Nitric Oxide Synthase|NOS Type 1|Brain NOS
- Storage buffer:saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Peptide sequence around aa.850~854 (F-N-S-V-S) derived from Human nNOS.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. no.:89304-846
- Supplier No.:GTX50497
Specifications
About this item
Rabbit Polyclonal antibody to nNOS
Suggested starting dilutions are as follows. WB: 1:500~1:1000, ICC/IF: 1:100~1:200. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: nNOS (nitric oxide Synthase 1 (neuronal))
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat