To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 27
- Clonality:Polyclonal
- Gene ID:246779
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:IL27
- Conjugation:Unconjugated
- ELISA:Yes
- Size:50 μg
- Western blot:Yes
- Antigen synonyms:IL27p28|IL-27A|IL27A|IL-27|IL30|p28
- Storage buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage temperature:Store vial at 4 °C for short-term use. For extended storage aliquot contents and freeze at –20 °C or below Avoid cycles of freezing and thawing.
- Concentration:1 mg/ml
- Immunogen:This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant mouse IL27/p28 protein.
- Purification:IgG fraction
- Cat. no.:89317-558
- Supplier No.:GTX48686
Specifications
About this item
Rabbit Polyclonal antibody to IL-27
Recommended Starting Dilutions: WB: Use at 1:500 - 1:2,000 ELISA: Use at 1:1,000 - 1:5,000
Type: Primary
Antigen: IL27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse