To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:heart and neural crest derivatives expressed transcript 2
- Clonality:Polyclonal
- Gene ID:58150
- Host:Goat
- Isotype:IgG
- Reactivity:Zebrafish
- Antigen symbol:HAND2
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5 mg/ml
- Immunogen:C-NDKKTKGRTGWPQH
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89293-148
- Supplier No.:GTX48501
Specifications
About this item
Goat polyclonal antibody to Hand2 (zebrafish)
Not yet tested. At this stage we are dependent on researchers in the field for further characterization of this product. Therefore we cannot recommend an optimal concentration and the product is investigative grade. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates?
Type: Primary
Antigen: hand2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Zebra Fish