To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Sid1 Transmembrane Family, Member 1
- Clonality:Polyclonal
- Gene ID:54847
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Antigen symbol:Cl channel interacting Protein (SID1 transmembrane Family member 1)
- Conjugation:Unconjugated
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:B830021E24Rik|SID-1|SID1
- Storage buffer:Phosphate/Glycine, pH7.2-7.4
- Storage temperature:Product should be aliquoted and stored at -20∞C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide
- Purification:Affinity purified using antigen.
- Cat. no.:89279-696
- Supplier No.:GTX47824
Specifications
About this item
Rabbit polyclonal antibody to Cl channel interacting protein
We recommend the following starting dilutions: 1:500 for WB 1:150 for IHC Optimal working dilutions should be determined experimentally be the end user.
Type: Primary
Antigen: Cl channel interacting Protein (SID1 transmembrane Family member 1)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: