To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:TIMELESS Interacting Protein
- Clonality:Polyclonal
- Gene ID:54962
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:TIPIN
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ImmunoFluorescence:Yes
- Size:50 μg
- Storage buffer:Phosphate, pH7.2-7.4
- Storage temperature:Product should be aliquoted and stored at -20∞C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide
- Purification:Affinity purified using antigen.
- Cat. no.:89275-830
- Supplier No.:GTX47795
Specifications
About this item
Rabbit Polyclonal antibody to TIPIN
We recommend the following starting dilutions: 1:500 for WB 1:150 for IHC Optimal working dilutions should be determined experimentally be the end user.
Type: Primary
Antigen: TIPIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse