To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Transmembrane Protease Serine 4
- Clonality:Polyclonal
- Gene ID:56649
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:TMPRSS4
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Epitope:245-257 aa
- Antigen synonyms:TMPRSS3|transmembrane serine protease 3|membrane-type serine protease 2|serine 4|MT-SP2|transmembrane protease|type II membrane serine protease
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5 mg/ml
- Immunogen:HCFRKHTDVFNWK
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89295-486
- Supplier No.:GTX47593
Specifications
About this item
Goat polyclonal antibody to TMPRSS4 (aa245-57)
Approx. 48kDa band observed in lysates of cell lines HEK293 and A549 (calculated MW of 48.2kDa according to NP_063947.1). Recommended concentration: 0.3-1µg/ml. This antibody is expected to recognise both reported isoforms (NP_063947.1 and NP_001077416.1).
Type: Primary
Antigen: TMPRSS4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: 245-257 aa
Host: Goat
Isotype:
Reactivity: Human