To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Inter-alpha (globulin) Inhibitor H4
- Clonality:Polyclonal
- Gene ID:3700
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ITIH4
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μg
- Antigen synonyms:ITIHL1|IHRP|GP120|PK120|PK-120|ITI-HC4|H4P
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5 mg/ml
- Immunogen:ERLCVDPRHRQGPVN
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89293-106
- Supplier No.:GTX47566
Specifications
About this item
Goat polyclonal antibody to ITIH4
Preliminary experiments gave an approx. 150kDa band in Human Liver and Lung lysates after 0.03µg/ml antibody staining. Please note that currently we cannot find an explanation in the literature for the band we observe given the calculated size of 103kDa according to NP_002209.2. The 150kDa band was successfully blocked by incubation with the immunizing peptide. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported?
Type: Primary
Antigen: ITIH4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human