To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Selectin P
- Clonality:Monoclonal
- Clone:Psel.KO.2.7
- Gene ID:281486
- Host:Mouse
- Isotype:IgG1
- Reactivity:Horse,Sheep,Goat,Human,Bovine,Rat,Mouse,Cat
- Antigen symbol:SELP
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:100 test
- Antigen synonyms:SELP|Lyam-1|Lnhr|Lyam1|PADGEM|antigen CD62,GRMP|LECAM3|CD62P|Ly-m22|Ly-22|LECAM-1|CD62L|AI528707|granule membrane protein 140kDa|PSEL|GMP140|L-selectin
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide 1% Bovine Serum Albumin 5% Sucrose
- Storage temperature:Store at 4°C. DO NOT FREEZE. This product should be stored undiluted. This product is photosensitive and should be protected from light. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. no.:89274-794
- Supplier No.:GTX43039
Specifications
About this item
Mouse monoclonal antibody [Psel.KO.2.7] to CD62P
Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: Psel.KO.2.7
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Cat Bovine, Goat, Horse, Rat, Sheep