Anti-ALPL Mouse Monoclonal Antibody [clone: BGN/3/661]
Catalog #: 89266-050
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alkaline Phosphatase liver/bone/kidney
- Clonality:Monoclonal
- Clone:BGN/03/661
- Gene ID:249
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:ALPL
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:TNSALP|AP-TNAP|TNAP|HOPS|APTNAP
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide (NaN3)
- Storage temperature:Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Alkaline phosphatase from human bone.
- Purification:Purified IgG prepared by affinity chromatography on Protein A
- Cat. no.:89266-050
- Supplier No.:GTX42810
Specifications
About this item
Mouse monoclonal antibody [BGN/03/661] to Alkaline Phosphatase
Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone: BGN/3/661
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Customers Who Bought This Also Bought