To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD200 Receptor 1
- Clonality:Monoclonal
- Clone:OX-110
- Gene ID:57781
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Antigen symbol:CD200R1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:100 μg
- Antigen synonyms:MOX2R|HCRTR2|CD200R|OX2R
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. no.:89292-666
- Supplier No.:GTX42286
Specifications
About this item
Rat monoclonal antibody [OX-110] to CD200R
Recommended Starting Dilutions: Flow Cytometry: 1/25 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: CD200R (CD200R 1)
Clonality: Monoclonal
Clone: OX-110
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse