To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SLACK (potassium channel subfamily T member 1)
- Clonality:Monoclonal
- Clone:S3-26
- Gene ID:60444
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Kcnt1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 μg
- Western blot:Yes
- Storage buffer:PBS pH7.4, 50% glycerol
- Storage temperature:Aliquot and store at -20 C. Avoid repeated freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Fusion protein amino acids 1168-1237 of rat Slo2.2 (Slack); NP_068625
- Purification:Protein G Purified
- Cat. no.:89265-960
- Supplier No.:GTX42002
Specifications
About this item
Mouse monoclonal antibody [S3-26] to SLACK
We recommend a working dilution of 1-10ug/mL (WB), 0.1-1.0ug/mL (Perox) (IHC/ICC), 1.0-10ug/mL (IF).
Type: Primary
Antigen: Kcnt1
Clonality: Monoclonal
Clone: S3-26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat