Order Entry
ContactUsLinkComponent
Anti-RANKL Rat Monoclonal Antibody [clone: IK22/5]
Anti-RANKL Rat Monoclonal Antibody [clone: IK22/5]
Catalog #: 89336-954
Supplier:  Genetex
Anti-RANKL Rat Monoclonal Antibody [clone: IK22/5]
Catalog #: 89336-954
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Receptor activator of nuclear factor kappa-B ligand
  • Clonality:
    Monoclonal
  • Clone:
    IK22/5
  • Gene ID:
    21943
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Antigen symbol:
    RANKL
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Size:
    100 μg
  • Western blot:
    Yes
  • Antigen synonyms:
    RANKLigand|Osteoprotegerin ligand|OPGL|sOdf|ODF|TRANCE|CD254|TNF-related activation-induced cytokine|RANKL|Receptor activator of nuclear factor kappa B ligand|hRANKL2|OPTB2
  • Storage buffer:
    Preservative: 0.09% Sodium Azide. Constituents: PBS. pH 7.2
  • Storage temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/ml
  • Immunogen:
    NSO derived recombinant mouse TRANCE.
  • Purification:
    Protein A purified
  • Cat. no.:
    89336-954
  • Supplier No.:
    GTX41426

Specifications

About this item

Rat monoclonal [IK22/5] to sRANKL

BL: Use at an assay dependent dilution. The Low Endotoxin Azide Free format is suggested for blocking studies. FACS: Use 0.5?g for 10^6 cells 100?l volume. IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 35 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RANKL
Clonality: Monoclonal
Clone: IK22/5
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Customers Who Bought This Also Bought