To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Immunoglobulin G1
- Clonality:Monoclonal
- Clone:B33/20
- Gene ID:3500
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:IgG1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:1 mg
- Storage buffer:Phosphate buffered saline, 0.09%Sodium Azide (NaN3)
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Purified human IgG
- Cat. no.:89280-376
- Supplier No.:GTX40390
Specifications
About this item
Mouse Monoclonal antibody [B33/20] to Human IgG
Optimal dilutions should be determined experimentally by the researcher.
Type: Secondary
Antigen: IgG1
Clonality: Monoclonal
Clone: B33/20
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human