Anti-ITGA9 Mouse Monoclonal Antibody [clone: Y9A2]
Catalog #: 89275-374
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 9
- Clonality:Monoclonal
- Clone:Y9A2
- Gene ID:3680
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Human,Guinea pig
- Antigen symbol:ITGA9
- Conjugation:Unconjugated
- Size:200 μg
- Antigen synonyms:ITGA4L|RLC|ALPHA-RLC
- Storage buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage temperature:Store at +4°C for one month or at -20°C for longer. This product should be stored undiluted. Avoid repeated freezing and thawing as this may denature the antibody. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. no.:89275-374
- Supplier No.:GTX39990
Specifications
About this item
Mouse monoclonal antibody [Y9A2] to Integrin alpha 9 + beta 1
Type: Primary
Antigen: ITGA9
Clonality: Monoclonal
Clone: Y9A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Customers Who Bought This Also Bought