Anti-ITGB5 Mouse Monoclonal Antibody [clone: P1F6]
Catalog #: 89262-382
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin Beta 5
- Clonality:Monoclonal
- Clone:P1F6
- Gene ID:3693
- Host:Mouse
- Isotype:IgG1
- Reactivity:Chicken,Human
- Antigen symbol:ITGB5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:50 μg
- Antigen synonyms:RGD1563276
- Storage buffer:Phosphate buffered saline containing 0.1% Sodium Azide (NaN3)
- Purification:Purified IgG prepared by affinity chromatography on Protein A from ascites
- Cat. no.:89262-382
- Supplier No.:GTX39987
Specifications
About this item
Mouse monoclonal antibody [P1F6] to Integrin alpha V + beta 5
Type: Primary
Antigen: ITGB5
Clonality: Monoclonal
Clone: P1F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Chicken
Customers Who Bought This Also Bought