To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:7,8-dihydro-8-oxoguanine triphosphatase
- Clonality:Polyclonal
- Gene ID:4521
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Antigen symbol:NUDT1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:200 μl
- Western blot:Yes
- Antigen synonyms:NUDIX Motif 1|MTH1|Nucleoside Diphosphate Linked Moiety X Motif 1|NUDT1
- Storage buffer:Phosphate-buffereed saline containing 0.05% Sodium Azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1.15 mg/ml
- Immunogen:A c-terminal peptide derived from the human MTH1.
- Purification:Antigen affinity purification
- Cat. no.:89264-800
- Supplier No.:GTX30108
Specifications
About this item
Rabbit polyclonal to MTH1
For WB: Use at a dilution of 1:500. Can be used for Western blot detection of MTH1 where a band is seen at ~ 18 kDa. For IHC-Fr: Use at a dilution of 1:200. Not tested in other applications. Optimal dilutions/concentrations shuld be determined by the researcher.
Type: Primary
Antigen: NUDT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat