Anti-CD20 Mouse Monoclonal Antibody [clone: MEM-97]
Catalog #: 89317-452
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD20 molecule
- Clonality:Monoclonal
- Clone:MEM-97
- Gene ID:931
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:CD20
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Membrane-spanning 4-domains subfamily A member 1|CVID5 MS4A2|MS4A1|Bp35|S7|LEU-16|CD20|B1
- Storage buffer:Phosphate buffered saline pH 7.4 containing 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Burkitt?s lymphoma cell line Raji.
- Purification:Protein A purified
- Cat. no.:89317-452
- Supplier No.:GTX28237
Specifications
About this item
Mouse monoclonal [MEM - 97] to CD20
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: MEM-97
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Customers Who Bought This Also Bought