To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Alpha 1, Skeletal Muscle
- Clonality:Monoclonal
- Clone:HHF35
- Gene ID:58
- Host:Mouse
- Isotype:IgG1
- Reactivity:Hamster,Rabbit,Monkey,Human,Rat
- Antigen symbol:ACTA1
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 μl
- Antigen synonyms:ACT1|alpha skeletal muscle Actin|ASMA|alpha Actin 1|NEM2|ACTA|NEM1
- Storage buffer:10mM PBS with 0.05% BSA and 0.05% Sodium azide
- Storage temperature:Keep as concentrated solution, aliquot and store at 4°C. Do not freeze.
- Immunogen:The SDS extracted protein fraction of human myocardium.
- Purification:Tissue culture supernatant
- Cat. no.:89318-066
- Supplier No.:GTX27813
Specifications
About this item
Mouse monoclonal antibody [HHF35] to Muscle Actin
Immunohistochemistry (1:50 to 1:60): formalin-fixed, paraffin-embedded tissue sections. Prolonged fixation in buffered formalin can destroy the epitope. Also works on frozen sections.
Type: Primary
Antigen: ACT1
Clonality: Monoclonal
Clone: HHF35
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Hamster, Monkey, Rabbit, Rat