To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Forkhead Box E1
- Clonality:Polyclonal
- Gene ID:2304
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:FOXE1
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μg
- Antigen synonyms:FKHL15|forkhead (Drosophila)-like 15|forkhead box E1 (thyroid transcription factor 2)|HFKH4|FOXE1|drosophila|FOXE2|homolog-like 15|forkhead box E1|HFKL5|Forkhead|TITF2|TTF-2|thyroid transcription factor-2|forkhead box E2|TTF2
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5 mg/ml
- Immunogen:C-AYPGGIDRFVSAM
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89279-004
- Supplier No.:GTX25080
Specifications
About this item
Goat polyclonal antibody to FOXE1 / TTF2
Preliminary experiments gave an approx 70kDa band in Human Thymus and Thyroid Gland lysates after 0.3µg/ml antibody staining. Please note that currently we cannot find an explanation in the literature for the band we observe given the calculated size of 38.1kDa according to NP_004464.2. The 65kDa band was successfully blocked by incubation with the immunizing peptide. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? In paraffin embedded Human Testis shows strong nuclear staining in developing sperm cells. Recommended concentration, 2-3µg/ml. This product has also been successfully used in Human Thyroid Gland: Zhang et al, Pathol Int. 2006 May;56(5):240-5. PMID: 16669872.. Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: FOXE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human