To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Myotubularin Related Protein 9
- Clonality:Polyclonal
- Gene ID:66036
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Dog,Rat
- Antigen symbol:MTMR9
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:100 μg
- Antigen synonyms:MTMR9|MTMR8|DKFZp434K171|DKFZP434K171|myotubularin related protein 8|myotubularin related protein 9|myotubularin-related protein 9 variant|MGC126672|myotubularin-related protein 9|LIP-STYX|C8orf9
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5 mg/ml
- Immunogen:C-AELETEDGMQESP
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89279-002
- Supplier No.:GTX25075
Specifications
About this item
Goat polyclonal antibody to MTMR9 (C Terminus)
No signal obtained yet but low background observed in Human Brain and Kidney lysates at up to 1µg/ml. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Immunohistochemsitry: This product has been successfully used on Rat Hypothalamus (Yanagiya et al, Hum Mol Genet. 2007 Dec15;16(24):3017-26. PMID: 17855449).
Type: Primary
Antigen: MTMR9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Dog, Rat