To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 2B
- Clonality:Monoclonal
- Clone:IVA30
- Gene ID:515011
- Host:Mouse
- Isotype:IgG2
- Reactivity:Bovine
- Antigen symbol:ITGA2B
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Size:100 μg
- Antigen synonyms:GP2B|GTA|platelet fibrinogen receptor|alpha subunit|CD41B|platelet membrane glycoprotein IIb|HPA3|integrin alpha-IIb|GPIIb|BDPLT2|CD41|GT|ITGA2B|platelet-specific antigen BAK|GPalpha IIb
- Storage buffer:Phosphate buffered saline pH 7.4 containing 15mM sodium azide
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Bovine trombocytes.
- Purification:>90% by SDS-PAGE
- Cat. no.:89337-192
- Supplier No.:GTX23919
Specifications
About this item
Mouse monoclonal [IVA30] to Integrin alpha 2 beta + Integrin beta 3
Not tested in applications other than those listed above. Optimal dilutions / concentrations should be determined by the end user.
Type: Primary
Antigen: ITGA2B
Clonality: Monoclonal
Clone: IVA30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Bovine