To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Myosin 7A
- Clonality:Polyclonal
- Gene ID:4647
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Antigen symbol:MYO7A
- Conjugation:Unconjugated
- Size:200 μl
- Western blot:Yes
- Antigen synonyms:NSRD2|MYOVIIA|DFNB2|DFNA11|MYU7A|MYO7A|Unconventional myosin-VIIa|USH1B
- Storage buffer:PBS, 1 mg/ml BSA, 0.05% sodium azide
- Storage temperature:Upon receipt - Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide: S(16)GQEFDVPIGAVVKLC(31).Immunizing peptide corresponds to amino acid residues 16-31 from mouse myosin VIIa. This sequence is completely conserved in mouse, porcine, and human myosin VIIa.
- Purification:Immunogen affinity purified
- Cat. no.:89264-756
- Supplier No.:GTX23481
Specifications
About this item
Rabbit polyclonal to Myosin VIIa
Recommended Starting Dilutions:ICC/IF: 1:50IHC: 1:50 - 1:500WB: 1:200Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: MYO7A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse