To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Linker for activation of T cells Family, member 2
- Clonality:Monoclonal
- Clone:NAP-03
- Gene ID:7462
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:NTAL
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:LAB
- Storage buffer:Phosphate buffered saline pH 7.4 containing 15mM sodium azide
- Storage temperature:Store at 2-8ºC. Do not freeze. Do not use after expiration date stamped on vial label.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide conjugated to KLH, corresponding to amino acids 196-212 of Human NTAL.
- Purification:>95% by SDS-PAGE
- Cat. no.:89317-378
- Supplier No.:GTX21901
Specifications
About this item
IP: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 30 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: NTAL
Clonality: Monoclonal
Clone: NAP-03
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human