To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD20 molecule
- Clonality:Polyclonal
- Gene ID:931
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CD20
- Conjugation:Unconjugated
- ELISA:Yes
- Size:100 μg
- Western blot:Yes
- Antigen synonyms:Membrane-spanning 4-domains subfamily A member 1|CVID5 MS4A2|MS4A1|Bp35|S7|LEU-16|CD20|B1
- Storage buffer:Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
- Storage temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:0.5 mg/ml
- Immunogen:CQDQESSPIENDSSP
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:89288-066
- Supplier No.:GTX21310
Specifications
About this item
Approx 35kDa band observed in Human Lymph lysates (calculated MW of 33.1kDa according to NP_068769.2). Recommended concentration: 0.003-0.01µg/ml. Please note that both reported variants (NP_068769.2 and NP_690605.1) encode the same protein.
Type: Primary
Antigen: CD20
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human