To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 7
- Clonality:Monoclonal
- Clone:542
- Gene ID:6354
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Antigen symbol:MCP3
- Conjugation:Unconjugated
- ELISA:Yes
- Size:250μg
- Western blot:Yes
- Antigen synonyms:MCP3|NC28|MGC138465|SCYA7|SCYA6|MCP-3|MARC|MGC138463|Chemokine (C-C motif) ligand 7|FIC
- Storage buffer:Preservative: None. Constituents: 0.01M PBS. pH 7.2
- Storage temperature:Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Recombinant fragment (Human).(M.W. 8.5 kDa containing 76 amino acid residues).
- Purification:Protein G purified
- Cat. no.:89328-346
- Supplier No.:GTX18693
Specifications
About this item
ELISA: Use at an assay dependent dilution. WB: Use at a concentration of 0.5 - 2 ?g/ml. Predicted molecular weight: 8.5 kDa. This antibody can be used as a capture antibody in sandwich ELISA applications for human MCP-3 detection in combination with a monoclonal tracer/detection antibody (HRP conjugated). Suggested Capture Coating Dose: 3?g/well; Substrate:TMB.A Western blot using an antibody concentration of 0.5-2?g/mL will allow visualization of 0.1?g/lane of human MCP-3. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: MCP3
Clonality: Monoclonal
Clone: 542
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human