Order Entry
ContactUsLinkComponent
Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT20]
Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT20]
Catalog #: 89329-716
Supplier:  Genetex
Anti-CD20 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: LT20]
Catalog #: 89329-716
Supplier:  Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD20 molecule
  • Clonality:
    Monoclonal
  • Clone:
    LT20
  • Gene ID:
    931
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Antigen symbol:
    CD20
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Size:
    100 test
  • Antigen synonyms:
    Membrane-spanning 4-domains subfamily A member 1|CVID5 MS4A2|MS4A1|Bp35|S7|LEU-16|CD20|B1
  • Storage buffer:
    Preservative: 15mM Sodium Azide. Constituents: PBS, BSA. pH 7.4
  • Storage temperature:
    Keep as concentrated solution, aliquot and store at 4°C. Do not freeze. Keep protected from prolonged exposure to light.
  • Immunogen:
    Normal lymphocytes from lymph node (Human).
  • Purification:
    Protein A purified
  • Cat. no.:
    89329-716
  • Supplier No.:
    GTX18268

Specifications

About this item

FACS: Use at a concentration of 200 ?l/ml whole blood. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: LT20
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Customers Who Bought This Also Bought